siRNA Details
| Gene Symbol: PLEKHG1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001029884.1 | NP_001025055.1 | 3 | 693 | ||
| XM_005267064.3 | XP_005267121.1 | 2 | 2772 | |||
| XM_005267065.3 | XP_005267122.1 | 4 | 719 | |||
| XM_005267066.2 | XP_005267123.1 | 2 | 1576 | |||
| XM_006715521.1 | XP_006715584.1 | 4 | 946 | |||
| XM_006715522.3 | XP_006715585.1 | 2 | 2813 | |||
| XM_011535981.2 | XP_011534283.1 | 2 | 3569 | |||
| XM_011535982.2 | XP_011534284.1 | 2 | 3569 | |||
| XM_011535983.2 | XP_011534285.1 | 3 | 1994 | |||
| XM_017011102.1 | XP_016866591.1 | 2 | 1578 | |||
| GenBank mRNA | AK056300.1 | - | 2 | 592 | ||
| AK293288.1 | - | 2 | 624 | |||
| AK294798.1 | - | 2 | 747 | |||
| AK307631.1 | - | 2 | 646 | |||
| BC140864.1 | - | 1 | 430 | 9021732 | ||
| BC151134.1 | - | 1 | 430 | 9057034 | ||

PLEKHG1
Genomic Map