siRNA Details
| Gene Symbol: IFT80 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001190241.1 | NP_001177170.1 | 21 | 2947 | ||
| NM_001190242.1 | NP_001177171.1 | 19 | 2581 | |||
| NM_020800.2 | NP_065851.1 | 20 | 2727 | |||
| GenBank mRNA | AB037795.1 | - | 19 | 2471 | ||
| AK055904.1 | - | 20 | 3513 | |||
| AK303410.1 | - | 20 | 2725 | |||
| AL133045.1 | - | 5 | 566 | |||
| BC042027.1 | - | 13 | 1572 | |||
| BC101494.1 | - | 20 | 2369 | 8069000 | ||
| BC113669.1 | - | 20 | 2369 | 8322721 | ||

IFT80
Genomic Map