siRNA Details
| Gene Symbol: ARRDC3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_020801.2 | NP_065852.1 | 6 | 1224 | ||
| GenBank mRNA | AB037797.1 | - | 6 | 1196 | ||
| AK291753.1 | - | 6 | 1199 | |||
| BC015928.2 | - | 6 | 1191 | 3908916 | ||
| BC053619.1 | - | 6 | 1196 | 6155815 | ||

ARRDC3
Genomic Map