siRNA Details
| Gene Symbol: FNIP2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001323916.1 | NP_001310845.1 | 17 | 3512 | ||
| NM_020840.2 | NP_065891.1 | 17 | 3507 | |||
| XM_005263156.1 | XP_005263213.1 | 18 | 3597 | |||
| XM_005263158.1 | XP_005263215.1 | 18 | 3397 | |||
| XM_011532154.2 | XP_011530456.1 | 18 | 4377 | |||
| XM_017008487.1 | XP_016863976.1 | 19 | 3546 | |||
| GenBank mRNA | AB040883.1 | - | 17 | 3364 | ||
| AK022968.1 | - | 5 | 1304 | |||

FNIP2
Genomic Map