siRNA Details
| Gene Symbol: PHF12 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001033561.1 | NP_001028733.1 | 9 | 1893 | ||
| NM_001290131.1 | NP_001277060.1 | 9 | 1893 | |||
| NM_020889.2 | NP_065940.1 | 9 | 1893 | |||
| XM_005258015.3 | XP_005258072.1 | 8 | 1182 | |||
| XM_005258016.3 | XP_005258073.1 | 8 | 1202 | |||
| XM_011525078.2 | XP_011523380.1 | 8 | 1494 | |||
| XM_011525079.2 | XP_011523381.1 | 9 | 1876 | |||
| XM_011525082.2 | XP_011523384.1 | 9 | 1876 | |||
| XM_011525083.2 | XP_011523385.1 | 9 | 1876 | |||
| XM_017024903.1 | XP_016880392.1 | 8 | 1941 | |||
| XM_017024904.1 | XP_016880393.1 | 8 | 1461 | |||
| GenBank mRNA | AB040956.1 | - | 4 | 414 | ||
| AK024290.1 | - | 2 | 219 | |||
| AK160370.1 | - | 9 | 3920 | |||
| AK291420.1 | - | 9 | 1365 | |||
| AK294056.1 | - | 8 | 1322 | |||
| AL161953.1 | - | 8 | 1780 | |||
| AY030283.1 | - | 9 | 1895 | |||
| BC001657.1 | - | 7 | 1110 | |||
| BC110882.1 | - | 9 | 1710 | 6157238 | ||
| BC121043.1 | - | 9 | 1395 | 40117907 | ||
| BC121044.1 | - | 9 | 1395 | 40117911 | ||

PHF12
Genomic Map