siRNA Details
| Gene Symbol: PHRF1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001286581.1 | NP_001273510.1 | 8 | 1026 | ||
| NM_001286582.1 | NP_001273511.1 | 8 | 1023 | |||
| NM_001286583.1 | NP_001273512.1 | 8 | 1014 | |||
| NM_020901.3 | NP_065952.2 | 8 | 1026 | |||
| XM_005253025.4 | XP_005253082.1 | 8 | 984 | |||
| XM_005253027.3 | XP_005253084.1 | 8 | 972 | |||
| XM_011520236.2 | XP_011518538.1 | 8 | 1026 | |||
| XM_011520237.2 | XP_011518539.1 | 7 | 867 | |||
| GenBank mRNA | AB040975.1 | - | 8 | 986 | ||
| BC112931.1 | - | 6 | 2771 | |||
| BC136615.1 | - | 8 | 951 | 9020605 | ||
| BC144293.1 | - | 8 | 951 | 9052813 | ||
| BC144294.1 | - | 8 | 939 | 9052814 | ||
| BC144295.1 | - | 8 | 948 | 9052815 | ||
| BC146771.1 | - | 8 | 986 | 9007218 | ||

PHRF1
Genomic Map