siRNA Details
| Gene Symbol: EPB41L5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001184937.1 | NP_001171866.1 | 12 | 1194 | ||
| NM_001184938.2 | NP_001171867.1 | 12 | 1484 | |||
| NM_001184939.1 | NP_001171868.1 | 12 | 1194 | |||
| NM_020909.3 | NP_065960.2 | 12 | 1194 | |||
| NR_135920.1 | 12 | 1194 | ||||
| XM_006712651.1 | XP_006712714.1 | 12 | 1174 | |||
| XM_006712652.1 | XP_006712715.1 | 12 | 1489 | |||
| XM_006712653.1 | XP_006712716.1 | 12 | 1194 | |||
| XM_011511529.2 | XP_011509831.1 | 12 | 1174 | |||
| XM_017004567.1 | XP_016860056.1 | 12 | 1030 | |||
| XM_017004568.1 | XP_016860057.1 | 12 | 1194 | |||
| GenBank mRNA | AB046768.1 | - | 5 | 443 | ||
| AK023019.1 | - | 12 | 1145 | |||
| AK290895.1 | - | 12 | 1104 | |||
| BC032822.2 | - | 12 | 1484 | 4827274 | ||
| BC054508.1 | - | 12 | 1129 | 5502663 | ||

EPB41L5
Genomic Map