siRNA Details
| Gene Symbol: CACHD1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001293274.1 | NP_001280203.1 | 25, 26 | 3807 | ||
| NM_020925.2 | NP_065976.2 | 26, 27 | 3528 | |||
| XM_011541862.1 | XP_011540164.1 | 25, 26 | 3803 | |||
| XM_011541863.1 | XP_011540165.1 | 26, 27 | 3952 | |||
| GenBank mRNA | AB046793.1 | - | 24, 25 | 3310 | ||
| AK022571.1 | - | 13, 14 | 1626 | |||
| AK024256.1 | - | 16, 17 | 1953 | |||
| AK299361.1 | - | 12, 13 | 1371 | |||
| AL833000.1 | - | 16, 17 | 1977 | |||
| BC039301.1 | - | 26, 27 | 3514 | 4836806 | ||
| BX537603.1 | - | 25, 26 | 3806 | |||

CACHD1
Genomic Map