siRNA Details
| Gene Symbol: WDFY4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_020945.1 | NP_065996.1 | 19 | 3620 | ||
| XM_005270004.3 | XP_005270061.1 | 19 | 3735 | |||
| XM_011539986.2 | XP_011538288.2 | 19 | 3738 | |||
| XM_011539987.2 | XP_011538289.2 | 19 | 3741 | |||
| XM_011539988.2 | XP_011538290.1 | 20 | 3819 | |||
| XM_011539990.2 | XP_011538292.1 | 19 | 3766 | |||
| XM_011539991.2 | XP_011538293.1 | 19 | 3770 | |||
| XM_011539992.2 | XP_011538294.1 | 19 | 3770 | |||
| XM_017016463.1 | XP_016871952.1 | 20 | 3662 | |||
| XM_017016465.1 | XP_016871954.1 | 19 | 3747 | |||
| GenBank mRNA | BM470200.1 | - | 4 | 680 | ||

WDFY4
Genomic Map