siRNA Details
| Gene Symbol: WDFY4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_020945.1 | NP_065996.1 | 50 | 7898 | ||
| XM_005270004.3 | XP_005270061.1 | 50 | 8013 | |||
| XM_011539986.2 | XP_011538288.2 | 50 | 8016 | |||
| XM_011539987.2 | XP_011538289.2 | 50 | 8019 | |||
| XM_011539988.2 | XP_011538290.1 | 51 | 8097 | |||
| XM_017016463.1 | XP_016871952.1 | 51 | 7940 | |||
| XM_017016464.1 | XP_016871953.1 | 32 | 4327 | |||
| GenBank mRNA | AB046827.1 | - | 18 | 2081 | ||
| AK024502.1 | - | 1 | 65 | |||
| AK074085.1 | - | 31 | 3956 | |||
| AK074443.1 | - | 13 | 1301 | |||
| AK301736.1 | - | 4 | 354 | |||
| AL833669.1 | - | 11 | 1027 | |||
| BC047574.1 | - | 11 | 967 | |||

WDFY4
Genomic Map