siRNA Details
| Gene Symbol: PTPN2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001207013.1 | NP_001193942.1 | 8 | 1068 | ||
| NM_001308287.1 | NP_001295216.1 | 6 | 894 | |||
| NM_002828.3 | NP_002819.2 | 7 | 999 | |||
| NM_080422.2 | NP_536347.1 | 7 | 999 | |||
| NM_080423.2 | NP_536348.1 | 7 | 999 | |||
| XM_005258124.3 | XP_005258181.1 | 8 | 1066 | |||
| XM_005258125.3 | XP_005258182.1 | 8 | 1066 | |||
| XM_011525705.2 | XP_011524007.1 | 9 | 1286 | |||
| XM_011525706.2 | XP_011524008.1 | 6 | 787 | |||
| XM_011525707.2 | XP_011524009.1 | 3 | 394 | |||
| XM_017025884.1 | XP_016881373.1 | 7 | 1015 | |||
| XM_017025885.1 | XP_016881374.1 | 8 | 1201 | |||
| XM_017025886.1 | XP_016881375.1 | 6 | 889 | |||
| XM_017025887.1 | XP_016881376.1 | 6 | 840 | |||
| XM_017025888.1 | XP_016881377.1 | 6 | 840 | |||
| GenBank mRNA | AB209569.1 | - | 6 | 926 | ||
| AK225706.1 | - | 8 | 947 | |||
| AK290649.1 | - | 7 | 881 | |||
| AK292570.1 | - | 7 | 899 | |||
| BC008244.1 | - | 7 | 842 | 3872164 | ||
| BC016727.1 | - | 7 | 911 | 4081330 | ||
| M25393.1 | - | 7 | 865 | |||

PTPN2
Genomic Map