siRNA Details
| Gene Symbol: WDR19 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001317924.1 | NP_001304853.1 | 32 | 3689 | ||
| NM_025132.3 | NP_079408.3 | 33 | 3805 | |||
| XM_011513724.1 | XP_011512026.1 | 33 | 3727 | |||
| XM_011513725.2 | XP_011512027.1 | 32 | 3682 | |||
| XM_011513726.2 | XP_011512028.1 | 32 | 3590 | |||
| XM_011513727.1 | XP_011512029.1 | 31 | 3510 | |||
| XM_017008501.1 | XP_016863990.1 | 31 | 3498 | |||
| GenBank mRNA | AB046858.1 | - | 23 | 2649 | ||
| AK122908.1 | - | 2 | 1962 | |||
| AK294730.1 | - | 32 | 3564 | |||
| AK296112.1 | - | 17 | 1779 | |||
| AK304161.1 | - | 32 | 3498 | |||
| AY029257.1 | - | 33 | 3805 | |||

WDR19
Genomic Map