siRNA Details
| Gene Symbol: G6PC2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001081686.1 | NP_001075155.1 | 1 | 253 | ||
| NM_021176.2 | NP_066999.1 | 1 | 253 | |||
| XM_011511564.2 | XP_011509866.1 | 1 | 270 | |||
| XM_011511565.2 | XP_011509867.1 | 1 | 26 | |||
| GenBank mRNA | BC104778.1 | - | 1 | 253 | 8143781 | |
| BC113376.1 | - | 1 | 253 | 8322428 | ||
| BQ268961.1 | - | 1 | 188 | |||
| CR627438.1 | - | 1 | 204 | |||

G6PC2
Genomic Map