siRNA Details
| Gene Symbol: CYP4F11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001128932.1 | NP_001122404.1 | 10 | 1226 | ||
| NM_021187.3 | NP_067010.3 | 9 | 1648 | |||
| GenBank mRNA | AF236085.1 | - | 9 | 1298 | ||
| AK125714.1 | - | 6 | 1353 | |||
| AK289424.1 | - | 9 | 1216 | |||
| AL833940.1 | - | 9 | 1198 | |||
| BC016853.1 | - | 10 | 1226 | 3846027 | ||

CYP4F11
Genomic Map