siRNA Details
| Gene Symbol: CYP4F11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001128932.1 | NP_001122404.1 | 3 | 271 | ||
| NM_021187.3 | NP_067010.3 | 2 | 693 | |||
| GenBank mRNA | AF236085.1 | - | 2 | 343 | ||
| AK289424.1 | - | 2 | 261 | |||
| AK309292.1 | - | 2 | 401 | |||
| AL833940.1 | - | 2 | 243 | |||
| BC016853.1 | - | 3 | 271 | 3846027 | ||
| BU180723.1 | - | 2 | 104 | |||

CYP4F11
Genomic Map