siRNA Details
| Gene Symbol: RPRD1B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_021215.3 | NP_067038.1 | 2, 3 | 691 | ||
| XM_005260477.3 | XP_005260534.1 | 2, 3 | 775 | |||
| XM_005260480.4 | XP_005260537.1 | 2, 3 | 775 | |||
| XM_011528954.2 | XP_011527256.1 | 2, 3 | 775 | |||
| XM_017027990.1 | XP_016883479.1 | 2, 3 | 775 | |||
| GenBank mRNA | AK312468.1 | - | 2, 3 | 451 | ||
| BC001696.2 | - | 2, 3 | 441 | |||
| BC033629.1 | - | 2, 3 | 475 | 5223673 | ||
| DQ372938.1 | - | 2, 3 | 564 | |||
More Information
Other Species Targeted
This siRNA also targets the following transcripts in Mouse: NM_001291134, NM_001291135, NM_027434, AK152220, AK147718, and others.

RPRD1B
Genomic Map