siRNA Details
| Gene Symbol: ABRACL | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_021243.2 | NP_067066.1 | 3 | 372 | ||
| XM_017011151.1 | XP_016866640.1 | 3 | 357 | |||
| GenBank mRNA | AF116682.1 | - | 3 | 307 | ||
| AF161398.1 | - | 3 | 353 | |||
| BC014953.1 | - | 3 | 245 | |||
| BG563686.1 | - | 3 | 373 | |||

ABRACL
Genomic Map