siRNA Details
| Gene Symbol: SNX6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_021249.3 | NP_067072.3 | 8 | 924 | ||
| NM_152233.2 | NP_689419.2 | 9 | 819 | |||
| XM_006720224.1 | XP_006720287.1 | 8 | 706 | |||
| GenBank mRNA | AF109364.1 | - | 4 | 302 | ||
| AF121856.1 | - | 9 | 810 | |||
| AK021456.1 | - | 4 | 306 | |||
| AK292250.1 | - | 9 | 802 | |||
| AK296213.1 | - | 10 | 947 | |||
| AK308464.1 | - | 10 | 947 | |||
| BC001798.1 | - | 8 | 924 | 3354467 | ||
| BI870378.1 | - | 3 | 200 | |||
| U83194.1 | - | 9 | 787 | |||

SNX6
Genomic Map