siRNA Details
| Gene Symbol: RAB1A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_004161.4 | NP_004152.1 | 6 | 990 | ||
| NM_015543.1 | NP_056358.1 | 4 | 762 | |||
| XM_005264468.1 | XP_005264525.1 | 5 | 699 | |||
| GenBank mRNA | AF498929.1 | - | 6 | 604 | ||
| AK055927.1 | - | 4 | 605 | |||
| AK303678.1 | - | 5 | 698 | |||
| AK315495.1 | - | 6 | 799 | |||
| AL050268.1 | - | 4 | 566 | |||
| BC000905.2 | - | 6 | 782 | 2900705 | ||
| BX571747.1 | - | 6 | 787 | |||
| CR533479.1 | - | 4 | 376 | |||
| CR536488.1 | - | 6 | 604 | |||
| M28209.1 | - | 6 | 654 | |||
| X13898.1 | - | 6 | 604 | |||

RAB1A
Genomic Map