siRNA Details
| Gene Symbol: RAB3A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_002866.4 | NP_002857.1 | 3 | 460 | ||
| XM_011528164.1 | XP_011526466.1 | 3 | 457 | |||
| GenBank mRNA | AF254795.1 | - | 3 | 420 | ||
| AK289559.1 | - | 3 | 438 | |||
| BC011782.2 | - | 3 | 425 | 3939176 | ||

RAB3A
Genomic Map