siRNA Details
| Gene Symbol: CARD18 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_021571.3 | NP_067546.1 | 1 | 178 | ||
| GenBank mRNA | AF208005.1 | - | 1 | 170 | ||
| AI189838.1 | - | 2 | 258 | |||
| AW295205.1 | - | 2 | 492 | |||
| AY358231.1 | - | 1 | 182 | |||
| BC131610.1 | - | 1 | 170 | 40115072 | ||

CARD18
Genomic Map