siRNA Details
| Gene Symbol: RBBP7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001198719.1 | NP_001185648.1 | 2 | 464 | ||
| NM_002893.3 | NP_002884.1 | 2 | 476 | |||
| GenBank mRNA | AK091911.1 | - | 2 | 461 | ||
| AK291567.1 | - | 2 | 220 | |||
| BC114500.1 | - | 2 | 374 | 40083852 | ||
| BC114501.1 | - | 2 | 374 | 40083851 | ||
| CR541701.1 | - | 2 | 118 | |||
| DA924163.1 | - | 2 | 464 | |||
| U35143.1 | - | 2 | 379 | |||
| X72841.1 | - | 2 | 403 | |||
More Information
Other Species Targeted
This siRNA also targets the following transcripts in Mouse: NM_009031, AK078349, AK136110, U35142, AK147062, and others.

RBBP7
Genomic Map