siRNA Details
| Gene Symbol: RBP3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_002900.2 | NP_002891.1 | 1 | 2003 | ||
| GenBank mRNA | AB593121.1 | - | 1 | 1995 | ||
| BC039844.1 | - | 1 | 1993 | 5578501 | ||
| J03912.1 | - | 1 | 1886 | |||
| M22453.1 | - | 1 | 2004 | |||

RBP3
Genomic Map