siRNA Details
| Gene Symbol: RGR | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001012720.1 | NP_001012738.1 | 3, 4 | 385 | ||
| NM_001012722.1 | NP_001012740.1 | 3, 4 | 385 | |||
| NM_002921.3 | NP_002912.2 | 3, 4 | 397 | |||
| GenBank mRNA | BC011349.1 | - | 3, 4 | 371 | 4157985 | |
| BG912392.1 | - | 3, 4 | 353 | |||
| BX641033.1 | - | 2 | 722 | |||
| KU178307.1 | - | 3, 4 | 359 | |||
| KU178308.1 | - | 3, 4 | 347 | |||
| U14910.1 | - | 3, 4 | 385 | |||
More Information
Set Membership: |
|

RGR
Genomic Map