siRNA Details
| Gene Symbol: RGS13 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_002927.4 | NP_002918.1 | 7 | 635 | ||
| NM_144766.2 | NP_658912.1 | 6 | 595 | |||
| GenBank mRNA | AF030107.1 | - | 7 | 643 | ||
| AF493935.1 | - | 4 | 340 | |||
| AY562947.1 | - | 4 | 340 | |||
| BC016667.1 | - | 6 | 561 | 4340668 | ||
| BC056866.1 | - | 7 | 634 | 4419604 | ||
| BT006929.1 | - | 4 | 340 | |||
| CR536532.1 | - | 4 | 340 | |||

RGS13
Genomic Map