siRNA Details
| Gene Symbol: C12orf10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_021640.3 | NP_067653.3 | 6 | 868 | ||
| GenBank mRNA | AA972697.1 | - | 2 | 346 | ||
| AF111805.1 | - | 4 | 460 | |||
| AF289485.1 | - | 6 | 836 | |||
| BC013956.2 | - | 5 | 655 | |||
| BC028904.1 | - | 5 | 772 | 4454943 | ||
| BC051871.1 | - | 6 | 827 | 5170816 | ||
| BM551107.1 | - | 2 | 174 | |||

C12orf10
Genomic Map