siRNA Details
| Gene Symbol: EDA2R | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001199687.2 | NP_001186616.1 | 4, 5 | 461 | ||
| NM_001242310.1 | NP_001229239.1 | 3, 4 | 363 | |||
| NM_001324206.1 | NP_001311135.1 | 4, 5 | 463 | |||
| NM_021783.4 | NP_068555.1 | 4, 5 | 463 | |||
| NR_136726.1 | 4, 5 | 461 | ||||
| NR_136727.1 | 4, 5 | 463 | ||||
| XM_011531000.1 | XP_011529302.1 | 4, 5 | 413 | |||
| XM_011531001.1 | XP_011529303.1 | 4, 5 | 461 | |||
| XM_011531002.1 | XP_011529304.1 | 4, 5 | 413 | |||
| XM_011531004.1 | XP_011529306.1 | 4, 5 | 413 | |||
| XM_011531006.2 | XP_011529308.1 | 4, 5 | 453 | |||
| XM_011531007.2 | XP_011529309.1 | 4, 5 | 453 | |||
| XM_017029704.1 | XP_016885193.1 | 4, 5 | 413 | |||
| XM_017029706.1 | XP_016885195.1 | 4, 5 | 413 | |||
| GenBank mRNA | AF298812.1 | - | 3, 4 | 353 | ||
| AK296264.1 | - | 4, 5 | 413 | |||
| AK314881.1 | - | 4, 5 | 429 | |||
| AY152724.1 | - | 3, 4 | 353 | |||
| AY358735.1 | - | 3, 4 | 356 | |||
| AY358736.1 | - | 3, 4 | 356 | |||
| BC034919.1 | - | 4, 5 | 410 | 5434545 | ||
| BP249106.1 | - | 4, 5 | 429 | |||
| DA215848.1 | - | 4, 5 | 462 | |||
| DA574050.1 | - | 4, 5 | 413 | |||

EDA2R
Genomic Map