siRNA Details
| Gene Symbol: NAA35 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001321881.1 | NP_001308810.1 | 22 | 2208 | ||
| NM_001321882.1 | NP_001308811.1 | 22 | 2426 | |||
| NM_024635.3 | NP_078911.3 | 22 | 2212 | |||
| XM_005252127.3 | XP_005252184.1 | 22 | 2283 | |||
| XM_011518903.2 | XP_011517205.1 | 19 | 2015 | |||
| XM_011518904.2 | XP_011517206.1 | 16 | 1701 | |||
| XM_017015012.1 | XP_016870501.1 | 23 | 2271 | |||
| GenBank mRNA | AK026296.1 | - | 6 | 547 | ||
| AK292239.1 | - | 22 | 2212 | |||
| BC117427.1 | - | 22 | 2133 | 40125978 | ||
| BC117429.1 | - | 22 | 2129 | 40125980 | ||

NAA35
Genomic Map