siRNA Details
| Gene Symbol: CYB5RL | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001031672.2 | NP_001026842.2 | 6 | 835 | ||
| GenBank mRNA | AK090456.1 | - | 6 | 2946 | ||
| BC012781.2 | - | 5 | 619 | |||
| BC049198.1 | - | 4 | 449 | |||
| BC064419.1 | - | 6 | 808 | |||
| CD515193.1 | - | 4 | 581 | |||

CYB5RL
Genomic Map