siRNA Details
| Gene Symbol: PAPPA2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_020318.2 | NP_064714.2 | 3 | 2168 | ||
| NM_021936.2 | NP_068755.2 | 3 | 2168 | |||
| XM_005245422.3 | XP_005245479.1 | 3 | 2215 | |||
| XM_011509857.1 | XP_011508159.1 | 4 | 2085 | |||
| XM_011509858.2 | XP_011508160.1 | 3 | 2428 | |||
| XM_017002023.1 | XP_016857512.1 | 4 | 2266 | |||
| XM_017002024.1 | XP_016857513.1 | 4 | 2313 | |||
| XM_017002025.1 | XP_016857514.1 | 3 | 2419 | |||
| GenBank mRNA | AF311940.1 | - | 2 | 1004 | ||
| AF342989.1 | - | 3 | 2168 | |||
| AF342990.1 | - | 3 | 2168 | |||
| AJ278348.1 | - | 2 | 967 | |||
| BC117193.1 | - | 2 | 1776 | |||
| BC117195.1 | - | 2 | 1777 | |||
| BC152552.1 | - | 3 | 2156 | |||

PAPPA2
Genomic Map