siRNA Details
| Gene Symbol: ROBO1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_002941.3 | NP_002932.1 | 3 | 365 | ||
| XM_011533976.1 | XP_011532278.1 | 3 | 454 | |||
| XM_011533977.2 | XP_011532279.1 | 3 | 307 | |||
| XM_011533978.1 | XP_011532280.1 | 3 | 454 | |||
| XM_011533979.1 | XP_011532281.1 | 3 | 454 | |||
| XM_011533980.1 | XP_011532282.1 | 3 | 454 | |||
| XM_017006984.1 | XP_016862473.1 | 3 | 454 | |||
| GenBank mRNA | AF040990.1 | - | 2 | 157 | ||
| AK297481.1 | - | 2 | 313 | |||
| DR003097.1 | - | 372 | ||||

ROBO1
Genomic Map