siRNA Details
| Gene Symbol: CTXN3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001048252.2 | NP_001041717.1 | 3 | 763 | ||
| NM_001127385.1 | NP_001120857.1 | 3 | 572 | |||
| GenBank mRNA | AB219764.1 | - | 3 | 740 | ||
| AB219832.1 | - | 3 | 538 | |||
| BC071790.1 | - | 3 | 536 | |||
| BC147012.1 | - | 1 | 215 | 9056940 | ||

CTXN3
Genomic Map