siRNA Details
Gene Symbol: RPL38 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000999.3 | NP_000990.1 | 3, 4 | 193 | ||
NM_001035258.1 | NP_001030335.1 | 3, 4 | 158 | |||
GenBank mRNA | AK312119.1 | - | 3, 4 | 161 | ||
BC000603.2 | - | 3, 4 | 130 | 3346447 | ||
BQ276548.1 | - | 3, 4 | 189 | |||
BU532782.1 | - | 3, 4 | 161 | |||
CB124619.1 | - | 3, 4 | 183 | |||
CB128529.1 | - | 3, 4 | 136 | |||
Z26876.1 | - | 3, 4 | 175 |
