siRNA Details
| Gene Symbol: MBOAT4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001100916.1 | NP_001094386.1 | 3 | 1117 | ||
| XM_017013726.1 | XP_016869215.1 | 4 | 1720 | |||
| GenBank mRNA | AF359269.1 | - | 1 | 737 | ||
| EU518495.2 | - | 3 | 1127 | |||

MBOAT4
Genomic Map