siRNA Details
| Gene Symbol: MBOAT4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001100916.1 | NP_001094386.1 | 3 | 1373 | ||
| XM_017013726.1 | XP_016869215.1 | 4 | 1976 | |||
| GenBank mRNA | AF359269.1 | - | 1 | 993 | ||
| EU518495.2 | - | 3 | 1383 | |||

MBOAT4
Genomic Map