siRNA Details
Gene Symbol: RTN1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_021136.2 | NP_066959.1 | 6 | 2336 | ||
NM_206852.2 | NP_996734.1 | 4 | 731 | |||
XM_017021578.1 | XP_016877067.1 | 4 | 620 | |||
GenBank mRNA | AK128113.1 | - | 1 | 2196 | ||
AK290534.1 | - | 4 | 731 | |||
BC000314.2 | - | 4 | 516 | |||
BC003003.1 | - | 4 | 516 | |||
BC090862.1 | - | 6 | 2251 | 6179803 | ||
BC111694.1 | - | 7 | 2026 | 40035553 | ||
BX449967.2 | - | 4 | 719 | |||
CR457030.1 | - | 4 | 423 | |||
L10333.1 | - | 6 | 2249 | |||
L10334.1 | - | 4 | 1157 | |||
L10335.1 | - | 4 | 451 |
