siRNA Details
| Gene Symbol: ATXN7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_000333.3 | NP_000324.1 | 5, 6 | 1040 | ||
| NM_001177387.1 | NP_001170858.1 | 4, 5 | 597 | |||
| GenBank mRNA | AF032103.1 | - | 3, 4 | 179 | ||
| AF032105.1 | - | 5, 6 | 1040 | |||
| AJ000517.1 | - | 5, 6 | 1048 | |||

ATXN7
Genomic Map