siRNA Details
| Gene Symbol: CCL16 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_004590.3 | NP_004581.1 | 2 | 193 | ||
| XM_005258020.4 | XP_005258077.1 | 4 | 475 | |||
| GenBank mRNA | AB007454.1 | - | 2 | 193 | ||
| AF039955.1 | - | 2 | 184 | |||
| AF055467.1 | - | 2 | 151 | |||
| BC096352.4 | - | 3 | 389 | |||
| BC099662.3 | - | 2 | 158 | 40009719 | ||
| BC099663.1 | - | 3 | 389 | |||
| BC099664.4 | - | 3 | 389 | |||
| BQ722834.1 | - | 1 | 70 | |||
| U91746.1 | - | 2 | 117 | |||

CCL16
Genomic Map