siRNA Details
| Gene Symbol: FAM204A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001134672.1 | NP_001128144.1 | 2 | 179 | ||
| NM_022063.2 | NP_071346.1 | 3 | 380 | |||
| XM_005270024.1 | XP_005270081.1 | 4 | 557 | |||
| GenBank mRNA | AK023250.1 | - | 3 | 359 | ||
| AK026630.1 | - | 3 | 373 | |||
| BC023577.2 | - | 3 | 363 | 4765095 | ||
| BM837313.1 | - | 2 | 172 | |||
| CR457304.1 | - | 1 | 113 | |||
| DA168924.1 | - | 3 | 384 | |||

FAM204A
Genomic Map