siRNA Details
| Gene Symbol: FAM204A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001134672.1 | NP_001128144.1 | 4, 5 | 425 | ||
| NM_022063.2 | NP_071346.1 | 5, 6 | 626 | |||
| XM_005270024.1 | XP_005270081.1 | 6, 7 | 803 | |||
| GenBank mRNA | AK023250.1 | - | 5, 6 | 605 | ||
| AK026630.1 | - | 5, 6 | 619 | |||
| BC023577.2 | - | 5, 6 | 609 | 4765095 | ||
| BM837313.1 | - | 4, 5 | 418 | |||
| CR457304.1 | - | 3, 4 | 359 | |||

FAM204A
Genomic Map