siRNA Details
| Gene Symbol: THADA | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001083953.1 | NP_001077422.1 | 13 | 2398 | ||
| NM_001271643.1 | NP_001258572.1 | 13 | 2398 | |||
| NM_001271644.1 | NP_001258573.1 | 13 | 2186 | |||
| NM_022065.4 | NP_071348.3 | 13 | 2186 | |||
| NR_073394.1 | 13 | 2178 | ||||
| XM_006712061.3 | XP_006712124.1 | 14 | 2233 | |||
| XM_006712062.1 | XP_006712125.1 | 13 | 2186 | |||
| XM_006712063.1 | XP_006712126.1 | 13 | 2066 | |||
| XM_006712064.2 | XP_006712127.1 | 13 | 2186 | |||
| XM_006712065.1 | XP_006712128.1 | 13 | 2186 | |||
| XM_006712066.1 | XP_006712129.1 | 13 | 2186 | |||
| XM_006712067.1 | XP_006712130.1 | 13 | 2186 | |||
| XM_006712068.3 | XP_006712131.1 | 13 | 2186 | |||
| XM_011533015.2 | XP_011531317.1 | 13 | 2186 | |||
| XM_017004674.1 | XP_016860163.1 | 13 | 2066 | |||
| XM_017004675.1 | XP_016860164.1 | 13 | 2149 | |||
| GenBank mRNA | AB051554.2 | - | 13 | 2398 | ||
| AK126004.1 | - | 13 | 2101 | |||
| AK126824.1 | - | 3 | 795 | |||
| AK290023.1 | - | 13 | 2186 | |||
| AL832141.1 | - | 13 | 2171 | |||
| AY149629.1 | - | 13 | 2178 | |||
| AY149633.1 | - | 13 | 2170 | |||
| BC037990.2 | - | 13 | 2163 | |||
| BX641038.1 | - | 14 | 2195 | |||
| BX648155.1 | - | 13 | 2182 | |||

THADA
Genomic Map