siRNA Details
| Gene Symbol: GPATCH3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_022078.2 | NP_071361.2 | 1 | 379 | ||
| GenBank mRNA | AK022517.1 | - | 1 | 374 | ||
| AK057930.1 | - | 1 | 374 | |||
| AK303183.1 | - | 1 | 376 | |||
| AL833886.1 | - | 1 | 361 | |||
| BC007767.2 | - | 1 | 355 | 4298740 | ||
| CR936767.1 | - | 1 | 379 | |||
| DA513389.1 | - | 1 | 379 | |||

GPATCH3
Genomic Map