siRNA Details
| Gene Symbol: GPATCH3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_022078.2 | NP_071361.2 | 2 | 819 | ||
| GenBank mRNA | AK022517.1 | - | 2 | 814 | ||
| AK057930.1 | - | 2 | 814 | |||
| AK303183.1 | - | 3 | 762 | |||
| AK308100.1 | - | 2 | 682 | |||
| AL833886.1 | - | 2 | 801 | |||
| BC007767.2 | - | 2 | 795 | 4298740 | ||
| CR936767.1 | - | 2 | 819 | |||

GPATCH3
Genomic Map