siRNA Details
Gene Symbol: CXorf56 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001170569.1 | NP_001164040.1 | 6 | 733 | ||
NM_001170570.1 | NP_001164041.1 | 5 | 609 | |||
NM_022101.3 | NP_071384.1 | 6 | 651 | |||
GenBank mRNA | AK096297.1 | - | 6 | 697 | ||
BC023506.2 | - | 6 | 612 | |||
BQ223940.1 | - | 6 | 616 |
Gene Symbol: LOC728024 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_003671.2 | 1 | 614 | |||
GenBank mRNA | BI758198.1 | - | 1 | 618 | ||
CR977513.1 | - | 1 | 521 |
