siRNA Details
| Gene Symbol: DMRTC2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001040283.2 | NP_001035373.1 | 6 | 771 | ||
| XM_011527214.1 | XP_011525516.1 | 6 | 783 | |||
| XM_011527215.1 | XP_011525517.1 | 6 | 775 | |||
| XM_017027123.1 | XP_016882612.1 | 6 | 788 | |||
| XM_017027124.1 | XP_016882613.1 | 6 | 790 | |||
| XM_017027125.1 | XP_016882614.1 | 6 | 793 | |||
| XM_017027126.1 | XP_016882615.1 | 6 | 883 | |||
| XM_017027127.1 | XP_016882616.1 | 6 | 748 | |||
| XM_017027128.1 | XP_016882617.1 | 6 | 953 | |||
| XM_017027129.1 | XP_016882618.1 | 6 | 784 | |||
| GenBank mRNA | AJ291669.1 | - | 5 | 587 | ||
| AK301822.1 | - | 6 | 774 | |||
| BC029202.1 | - | 6 | 737 | 5170924 | ||
| BC039266.1 | - | 6 | 739 | 5298983 | ||

DMRTC2
Genomic Map