siRNA Details
| Gene Symbol: SECTM1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_003004.2 | NP_002995.1 | 3 | 733 | ||
| XM_005256392.3 | XP_005256449.1 | 3 | 517 | |||
| XM_011523588.2 | XP_011521890.1 | 3 | 764 | |||
| GenBank mRNA | AK223068.1 | - | 3 | 426 | ||
| AK223104.1 | - | 3 | 457 | |||
| AK290708.1 | - | 3 | 686 | |||
| AK312980.1 | - | 3 | 453 | |||
| BC017716.1 | - | 3 | 427 | 4413387 | ||
| BI861812.1 | - | 1 | 154 | |||
| U77643.1 | - | 3 | 453 | |||

SECTM1
Genomic Map