siRNA Details
Gene Symbol: SNX16 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_022133.3 | NP_071416.2 | 7, 8 | 1333 | ||
NM_152836.2 | NP_690049.1 | 6, 7 | 1087 | |||
NM_152837.2 | NP_690050.1 | 5, 6 | 1000 | |||
XM_005251282.4 | XP_005251339.1 | 7, 8 | 1040 | |||
XM_005251283.2 | XP_005251340.1 | 8, 9 | 1177 | |||
XM_011517574.2 | XP_011515876.1 | 5, 6 | 430 | |||
GenBank mRNA | AF305779.1 | - | 6, 7 | 938 | ||
AK290903.1 | - | 6, 7 | 1003 | |||
AL833763.1 | - | 5, 6 | 907 | |||
BC033630.1 | - | 6, 7 | 995 | 5224216 |
