siRNA Details
| Gene Symbol: POPDC2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001308333.1 | NP_001295262.1 | 1 | 191 | ||
| NM_022135.3 | NP_071418.2 | 1 | 191 | |||
| XM_005247698.3 | XP_005247755.1 | 1 | 191 | |||
| GenBank mRNA | AF204173.1 | - | 1 | 174 | ||
| AK314819.1 | - | 1 | 192 | |||
| BC044929.1 | - | 1 | 191 | 5191230 | ||

POPDC2
Genomic Map