siRNA Details
| Gene Symbol: CRLF2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001012288.2 | NP_001012288.2 | 7 | 974 | ||
| NM_022148.3 | NP_071431.2 | 8 | 1077 | |||
| NR_110830.1 | 8 | 1256 | ||||
| XM_011546181.2 | XP_011544483.1 | 8 | 1112 | |||
| GenBank mRNA | AB052639.1 | - | 8 | 988 | ||
| AF142570.1 | - | 8 | 1004 | |||
| AF338733.1 | - | 8 | 988 | |||
| AK026800.1 | - | 8 | 1258 | |||
| AK223303.1 | - | 8 | 1104 | |||
| AK225583.1 | - | 8 | 1258 | |||
| AY775789.1 | - | 7 | 885 | |||

CRLF2
Genomic Map