siRNA Details
| Gene Symbol: ACOT6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001037162.1 | NP_001032239.1 | 2 | 655 | ||
| XM_011537076.2 | XP_011535378.1 | 3 | 1262 | |||
| GenBank mRNA | BC126378.1 | - | 2 | 422 | 8992094 | |
| BC126380.1 | - | 2 | 422 | 8992096 | ||
| DQ082756.1 | - | 2 | 655 | |||

ACOT6
Genomic Map